siRNA Details
Gene Symbol: S1pr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010101.4 | NP_034231.1 | 2 | 939 | ||
GenBank mRNA | AB028143.1 | - | 2 | 807 | ||
AK028043.1 | - | 2 | 796 | |||
AK029852.1 | - | 2 | 795 | |||
AK030134.1 | - | 2 | 794 | |||
AK047268.1 | - | 2 | 787 | |||
AK078443.1 | - | 2 | 793 | |||
AK081919.1 | - | 2 | 784 | |||
AK083405.1 | - | 2 | 772 | |||
AK084944.1 | - | 2 | 794 | |||
AK085180.1 | - | 2 | 794 | |||
AK087076.1 | - | 2 | 794 | |||
AK087139.1 | - | 2 | 793 | |||
BC064007.1 | - | 2 | 779 | 30244496 | ||
BC068176.1 | - | 2 | 779 | 30613259 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
