siRNA Details
Gene Symbol: Egr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010118.3 | NP_034248.2 | 2 | 818 | ||
GenBank mRNA | AK153882.1 | - | 2 | 818 | ||
AK155585.1 | - | 2 | 819 | |||
BC009093.1 | - | 2 | 583 | 3157863 | ||
X06746.1 | - | 3 | 708 |
