siRNA Details
| Gene Symbol: Egr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010118.3 | NP_034248.2 | 2 | 1549 | ||
| GenBank mRNA | AK153882.1 | - | 2 | 1549 | ||
| AK155585.1 | - | 2 | 1550 | |||
| BC009093.1 | - | 2 | 1314 | 3157863 | ||
| X06746.1 | - | 3 | 1439 | |||

Egr2
Genomic Map