siRNA Details
| Gene Symbol: Emb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010330.4 | NP_034460.3 | 5, 6 | 892 | ||
| GenBank mRNA | AK079570.1 | - | 5, 6 | 834 | ||
| AK088480.1 | - | 5, 6 | 720 | |||
| AK138271.1 | - | 5, 6 | 841 | |||
| AK148449.1 | - | 5, 6 | 720 | |||
| AK148540.1 | - | 5, 6 | 720 | |||
| AK150625.1 | - | 5, 6 | 721 | |||
| AK161679.1 | - | 5, 6 | 720 | |||
| AK166602.1 | - | 5, 6 | 720 | |||
| AK167997.1 | - | 5, 6 | 752 | |||
| BC014858.1 | - | 5, 6 | 711 | 4500922 | ||
| J03535.2 | - | 5, 6 | 719 | |||

Emb
Genomic Map