siRNA Details
| Gene Symbol: Evx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007966.4 | NP_031992.1 | 1 | 411 | ||
| GenBank mRNA | BC126907.1 | - | 1 | 169 | 40059181 | |
| X54239.1 | - | 1 | 412 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Evx1| Gene Symbol: Evx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007966.4 | NP_031992.1 | 1 | 411 | ||
| GenBank mRNA | BC126907.1 | - | 1 | 169 | 40059181 | |
| X54239.1 | - | 1 | 412 | |||