siRNA Details
| Gene Symbol: Kcnn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080466.2 | NP_536714.2 | 4 | 1776 | ||
| GenBank mRNA | AF357241.1 | - | 4 | 1516 | ||
| AK137738.1 | - | 4 | 1777 | |||
| BC106769.1 | - | 4 | 1518 | 40039722 | ||
| BC106770.1 | - | 4 | 1518 | 40039723 | ||

Kcnn3
Genomic Map