siRNA Details
| Gene Symbol: Fas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007987.2 | NP_032013.2 | 9 | 742 | ||
| GenBank mRNA | AK002590.1 | - | 9 | 693 | ||
| BC061160.1 | - | 9 | 715 | 30302649 | ||
| DQ846748.1 | - | 9 | 686 | |||
| M83649.1 | - | 9 | 735 | |||

Fas
Genomic Map