siRNA Details
| Gene Symbol: Ms4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013516.2 | NP_038544.1 | 5 | 553 | ||
| GenBank mRNA | BC139323.1 | - | 5 | 477 | 8862345 | |
| BC139324.1 | - | 5 | 477 | 8862346 | ||
| J05019.1 | - | 5 | 477 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ms4a2| Gene Symbol: Ms4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013516.2 | NP_038544.1 | 5 | 553 | ||
| GenBank mRNA | BC139323.1 | - | 5 | 477 | 8862345 | |
| BC139324.1 | - | 5 | 477 | 8862346 | ||
| J05019.1 | - | 5 | 477 | |||