siRNA Details
| Gene Symbol: Fcna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007995.3 | NP_032021.1 | 1, 2 | 219 | ||
| GenBank mRNA | AB007813.1 | - | 1, 2 | 111 | ||
| AB222271.1 | - | 1, 2 | 21 | |||
| AK133539.1 | - | 1, 2 | 196 | |||
| AK138752.1 | - | 1, 2 | 166 | |||
| AK159651.1 | - | 1, 2 | 189 | |||
| BC019180.1 | - | 1, 2 | 167 | 5008934 | ||
| CT010383.1 | - | 1, 2 | 81 | |||

Fcna
Genomic Map