siRNA Details
| Gene Symbol: Fgf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010197.3 | NP_034327.1 | 3 | 387 | ||
| GenBank mRNA | AK035330.1 | - | 3 | 388 | ||
| AK142163.1 | - | 4 | 464 | |||
| BC027001.1 | - | 3 | 376 | |||
| BC037601.1 | - | 3 | 406 | 5137246 | ||
| M30641.1 | - | 2 | 200 | |||
| U67610.1 | - | 3 | 353 | |||

Fgf1
Genomic Map