siRNA Details
Gene Symbol: Fgfr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008011.2 | NP_032037.2 | 16, 17 | 2306 | ||
GenBank mRNA | AK084850.1 | - | 16, 17 | 2306 | ||
AK085723.1 | - | 16, 17 | 2304 | |||
BC033313.1 | - | 16, 17 | 2493 | 5132831 | ||
DQ388428.1 | - | 15, 16 | 2192 | |||
X59927.1 | - | 16, 17 | 2275 |
