siRNA Details
| Gene Symbol: Fgfr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008011.2 | NP_032037.2 | 15, 16 | 2162 | ||
| GenBank mRNA | AK084850.1 | - | 15, 16 | 2162 | ||
| BC033313.1 | - | 15, 16 | 2349 | 5132831 | ||
| DQ388428.1 | - | 14, 15 | 2048 | |||
| X59927.1 | - | 15, 16 | 2131 | |||

Fgfr4
Genomic Map