siRNA Details
| Gene Symbol: Fjx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010218.2 | NP_034348.2 | 1 | 1209 | ||
| GenBank mRNA | AK051097.1 | - | 1 | 1186 | ||
| BC051990.1 | - | 1 | 1209 | 5687527 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fjx1| Gene Symbol: Fjx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010218.2 | NP_034348.2 | 1 | 1209 | ||
| GenBank mRNA | AK051097.1 | - | 1 | 1186 | ||
| BC051990.1 | - | 1 | 1209 | 5687527 | ||