siRNA Details
| Gene Symbol: Fkbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010222.2 | NP_034352.1 | 1 | 254 | ||
| GenBank mRNA | AF040252.1 | - | 1 | 226 | ||
| AK042208.1 | - | 1 | 242 | |||
| AK138924.1 | - | 1 | 170 | |||
| BC032961.1 | - | 1 | 211 | 1347695 | ||

Fkbp7
Genomic Map