siRNA Details
| Gene Symbol: Fkbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010222.2 | NP_034352.1 | 3 | 470 | ||
| GenBank mRNA | AF040252.1 | - | 3 | 442 | ||
| AK138924.1 | - | 3 | 386 | |||
| BC032961.1 | - | 3 | 427 | 1347695 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fkbp7| Gene Symbol: Fkbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010222.2 | NP_034352.1 | 3 | 470 | ||
| GenBank mRNA | AF040252.1 | - | 3 | 442 | ||
| AK138924.1 | - | 3 | 386 | |||
| BC032961.1 | - | 3 | 427 | 1347695 | ||