siRNA Details
| Gene Symbol: Aktip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017861.1 | - | 6 | 599 | ||
| AK146459.1 | - | 7 | 715 | |||
| AK158704.1 | - | 7 | 717 | |||
| BC008130.1 | - | 8 | 720 | 3498984 | ||
| BC034861.1 | - | 7 | 724 | |||
| X71978.1 | - | 7 | 739 | |||
| Z67963.1 | - | 7 | 739 | |||

Aktip
Genomic Map