siRNA Details
| Gene Symbol: Aktip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017861.1 | - | 4, 5 | 490 | ||
| AK146459.1 | - | 5, 6 | 606 | |||
| AK158704.1 | - | 5, 6 | 608 | |||
| BC008130.1 | - | 6, 7 | 611 | 3498984 | ||
| BC034861.1 | - | 5, 6 | 615 | |||
| X71978.1 | - | 5, 6 | 630 | |||
| Z67963.1 | - | 5, 6 | 630 | |||

Aktip
Genomic Map