siRNA Details
| Gene Symbol: Gata5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008093.2 | NP_032119.2 | 1 | 746 | ||
| GenBank mRNA | AK132020.1 | - | 1 | 767 | ||
| AK142213.1 | - | 1 | 748 | |||
| BC105653.1 | - | 2 | 459 | 40046623 | ||
| BC105654.1 | - | 2 | 520 | 40046622 | ||
| U84725.1 | - | 1 | 740 | |||

Gata5
Genomic Map