siRNA Details
| Gene Symbol: Gbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010260.1 | NP_034390.1 | 11 | 1860 | ||
| GenBank mRNA | AF077007.1 | - | 11 | 1860 | ||
| AF109168.1 | - | 11 | 1813 | |||
| AJ007970.1 | - | 11 | 1853 | |||
| BC011336.1 | - | 11 | 1849 | 3494311 | ||
| BC032882.1 | - | 11 | 1875 | 1230883 | ||
| BC049141.1 | - | 11 | 1837 | |||
| CT010249.1 | - | 10 | 1690 | |||

Gbp2
Genomic Map