siRNA Details
| Gene Symbol: Gja5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271628.1 | NP_001258557.1 | 3 | 346 | ||
| NM_008121.3 | NP_032147.1 | 2 | 289 | |||
| GenBank mRNA | AK038780.1 | - | 1018 | |||
| AK137069.1 | - | 2 | 250 | |||
| AY390401.1 | - | 3 | 346 | |||
| BC053054.1 | - | 2 | 275 | 6401526 | ||

Gja5
Genomic Map