siRNA Details
| Gene Symbol: Gja5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271628.1 | NP_001258557.1 | 3 | 756 | ||
| NM_008121.3 | NP_032147.1 | 2 | 699 | |||
| GenBank mRNA | AK038780.1 | - | 1428 | |||
| AK137069.1 | - | 2 | 661 | |||
| BC053054.1 | - | 2 | 685 | 6401526 | ||

Gja5
Genomic Map