siRNA Details
| Gene Symbol: Gsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080552.1 | NP_001074021.1 | 3 | 426 | ||
| NM_001080553.1 | NP_001074022.1 | 3 | 579 | |||
| NM_010352.2 | NP_034482.2 | 4 | 639 | |||
| GenBank mRNA | AK006326.1 | - | 1 | 22 | ||
| AK088285.1 | - | 1 | 22 | |||
| BC023009.1 | - | 3 | 511 | 5364798 | ||
| D87325.1 | - | 4 | 518 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001013166, BC079000.

Gsg1
Genomic Map