siRNA Details
Gene Symbol: Gsx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133256.2 | NP_573555.1 | 1 | 180 | ||
GenBank mRNA | BC119464.1 | - | 1 | 179 | ||
BC119465.1 | - | 1 | 179 | |||
BC134359.1 | - | 1 | 179 | |||
S79041.1 | - | 1 | 180 |
