siRNA Details
| Gene Symbol: H13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159551.1 | NP_001153023.1 | 13 | 1390 | ||
| NM_010376.4 | NP_034506.1 | 12 | 1190 | |||
| GenBank mRNA | AK007664.1 | - | 12 | 1172 | ||
| AK012600.1 | - | 12 | 1183 | |||
| AK028109.1 | - | 12 | 1159 | |||
| AK049101.1 | - | 13 | 1364 | |||
| AK050664.1 | - | 12 | 1133 | |||
| AK077983.1 | - | 5 | 285 | |||
| AK080966.1 | - | 12 | 1152 | |||
| AK131644.1 | - | 1 | 185 | |||
| AK169922.1 | - | 12 | 1147 | |||
| AK171790.1 | - | 11 | 993 | |||
| DQ168449.1 | - | 13 | 1406 | |||

H13
Genomic Map