siRNA Details
| Gene Symbol: H2-Aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010378.2 | NP_034508.2 | 2 | 184 | ||
| GenBank mRNA | AF119253.1 | - | 2 | 173 | ||
| AF119254.1 | - | 2 | 173 | |||
| AK002532.1 | - | 2 | 235 | |||
| AK154000.1 | - | 2 | 190 | |||
| AK170844.1 | - | 2 | 2686 | |||
| AK170931.1 | - | 2 | 236 | |||
| AK171383.1 | - | 2 | 228 | |||
| AY452201.1 | - | 2 | 179 | |||
| BC019721.1 | - | 2 | 184 | 3669693 | ||
| BC031711.1 | - | 2 | 190 | 3670758 | ||
| K01922.1 | - | 2 | 156 | |||
| K01923.1 | - | 2 | 202 | |||
| K01926.1 | - | 1 | 92 | |||
| M11357.1 | - | 1 | 110 | |||
| M21931.1 | - | 2 | 202 | |||
| V00832.1 | - | 2 | 202 | |||
| X52643.1 | - | 2 | 173 | |||

H2-Aa
Genomic Map