siRNA Details
| Gene Symbol: H2-Aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010378.2 | NP_034508.2 | 2 | 307 | ||
| GenBank mRNA | AK002532.1 | - | 2 | 358 | ||
| BC019721.1 | - | 2 | 307 | 3669693 | ||
| BC031711.1 | - | 2 | 313 | 3670758 | ||
| K01922.1 | - | 2 | 279 | |||
| K01926.1 | - | 1 | 215 | |||

H2-Aa
Genomic Map