siRNA Details
| Gene Symbol: Hccs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008222.4 | NP_032248.3 | 4 | 571 | ||
| GenBank mRNA | AK077566.1 | - | 4 | 509 | ||
| AK143150.1 | - | 4 | 577 | |||
| BC061466.1 | - | 4 | 661 | 6813540 | ||
| BC062905.1 | - | 4 | 661 | 6813540 | ||
| U36788.1 | - | 3 | 356 | |||

Hccs
Genomic Map