siRNA Details
| Gene Symbol: Hoxa13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008264.1 | NP_032290.1 | 2 | 1044 | ||
| GenBank mRNA | BC100731.1 | - | 3 | 513 | 40045367 | |
| BC100732.1 | - | 3 | 510 | 40045368 | ||
| BC100733.1 | - | 3 | 510 | 40045370 | ||

Hoxa13
Genomic Map