siRNA Details
| Gene Symbol: Hprt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013556.2 | NP_038584.2 | 8 | 705 | ||
| GenBank mRNA | AK002286.1 | - | 8 | 650 | ||
| AK088114.1 | - | 8 | 595 | |||
| AK146626.1 | - | 8 | 651 | |||
| AY423851.1 | - | 2 | 60 | |||
| BC004686.1 | - | 8 | 620 | 3500885 | ||
| BC083145.1 | - | 8 | 591 | 30445781 | ||
| J00423.1 | - | 8 | 646 | |||

Hprt
Genomic Map