siRNA Details
Gene Symbol: Hras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130443.1 | NP_001123915.1 | 2 | 288 | ||
NM_001130444.1 | NP_001123916.1 | 2 | 288 | |||
NM_008284.2 | NP_032310.2 | 2 | 288 | |||
GenBank mRNA | AK183914.1 | - | 1 | 47 | ||
AY373386.1 | - | 1 | 82 | |||
BC011083.1 | - | 2 | 166 | 3152667 | ||
BC050837.1 | - | 2 | 278 | |||
BC061885.1 | - | 244 | ||||
BQ963436.1 | - | 2 | 277 | |||
CK788792.1 | - | 2 | 288 |
