siRNA Details
Gene Symbol: Hras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130443.1 | NP_001123915.1 | 2, 3 | 318 | ||
NM_001130444.1 | NP_001123916.1 | 2, 3 | 318 | |||
NM_008284.2 | NP_032310.2 | 2, 3 | 318 | |||
GenBank mRNA | AY373386.1 | - | 1, 2 | 112 | ||
BC011083.1 | - | 2, 3 | 196 | 3152667 | ||
BC050837.1 | - | 2, 3 | 308 | |||
BC061885.1 | - | 274 | ||||
BQ963436.1 | - | 2, 3 | 307 | |||
CK788792.1 | - | 2, 3 | 318 |
