siRNA Details
| Gene Symbol: Hsd17b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008291.3 | NP_032317.2 | 9 | 702 | ||
| GenBank mRNA | U66827.1 | - | 9 | 687 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hsd17b3| Gene Symbol: Hsd17b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008291.3 | NP_032317.2 | 9 | 702 | ||
| GenBank mRNA | U66827.1 | - | 9 | 687 | ||