siRNA Details
| Gene Symbol: Hsd17b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008291.3 | NP_032317.2 | 10, 11 | 871 | ||
| GenBank mRNA | BB041573.2 | - | 2, 3 | 252 | ||
| U66827.1 | - | 10, 11 | 856 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hsd17b3| Gene Symbol: Hsd17b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008291.3 | NP_032317.2 | 10, 11 | 871 | ||
| GenBank mRNA | BB041573.2 | - | 2, 3 | 252 | ||
| U66827.1 | - | 10, 11 | 856 | |||