siRNA Details
| Gene Symbol: Sdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008304.2 | NP_032330.1 | 3 | 725 | ||
| GenBank mRNA | AK011042.1 | - | 3 | 574 | ||
| AK204318.1 | - | 3 | 152 | |||
| AK216657.1 | - | 2 | 152 | |||
| BC003929.1 | - | 2 | 123 | |||
| BC047144.1 | - | 3 | 591 | 6395969 | ||
| U00674.1 | - | 3 | 725 | |||

Sdc2
Genomic Map