siRNA Details
| Gene Symbol: Sdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008304.2 | NP_032330.1 | 4, 5 | 962 | ||
| GenBank mRNA | AK011042.1 | - | 4, 5 | 811 | ||
| BC003929.1 | - | 3, 4 | 360 | |||
| BC047144.1 | - | 4, 5 | 828 | 6395969 | ||
| U00674.1 | - | 4, 5 | 962 | |||
| Gene Symbol: Dsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC026631.1 | - | 987 | |||

Sdc2
Genomic Map