siRNA Details
| Gene Symbol: Sdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008304.2 | NP_032330.1 | 4 | 866 | ||
| GenBank mRNA | AK011042.1 | - | 4 | 715 | ||
| BC047144.1 | - | 4 | 732 | 6395969 | ||
| U00674.1 | - | 4 | 866 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sdc2| Gene Symbol: Sdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008304.2 | NP_032330.1 | 4 | 866 | ||
| GenBank mRNA | AK011042.1 | - | 4 | 715 | ||
| BC047144.1 | - | 4 | 732 | 6395969 | ||
| U00674.1 | - | 4 | 866 | |||