siRNA Details
| Gene Symbol: Htr5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010483.3 | NP_034613.2 | 2 | 1183 | ||
| GenBank mRNA | BC114988.1 | - | 2 | 1072 | 40056207 | |
| BC115825.1 | - | 2 | 1072 | 40056204 | ||
| X69867.1 | - | 2 | 1218 | |||
More Information
Set Membership: |
|

Htr5b
Genomic Map