siRNA Details
| Gene Symbol: Irf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018533.1 | - | 7 | 671 | ||
| AK036488.1 | - | 7 | 671 | |||
| AK137450.1 | - | 7 | 1206 | |||
| AK146004.1 | - | 7 | 671 | |||
| AK150320.1 | - | 7 | 672 | |||
| AK150370.1 | - | 7 | 671 | |||
| AK151324.1 | - | 7 | 671 | |||
| AK151480.1 | - | 7 | 671 | |||
| AK152477.1 | - | 7 | 671 | |||
| AK152906.1 | - | 7 | 671 | |||
| AK153068.1 | - | 7 | 671 | |||
| AK153166.1 | - | 7 | 671 | |||
| AK153395.1 | - | 7 | 671 | |||
| AK154662.1 | - | 7 | 671 | |||
| AK156481.1 | - | 7 | 671 | |||
| AK157147.1 | - | 7 | 671 | |||
| AK157170.1 | - | 7 | 671 | |||
| AK157249.1 | - | 7 | 671 | |||
| AK157700.1 | - | 7 | 670 | |||
| AK159835.1 | - | 7 | 671 | |||
| BC005450.1 | - | 7 | 652 | 3487214 | ||
| CT010225.1 | - | 6 | 617 | |||
| M32489.1 | - | 7 | 637 | |||

Irf8
Genomic Map