siRNA Details
| Gene Symbol: Irf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018533.1 | - | 4, 5 | 504 | ||
| AK036488.1 | - | 4, 5 | 504 | |||
| AK137450.1 | - | 4, 5 | 1039 | |||
| AK146004.1 | - | 4, 5 | 504 | |||
| AK150320.1 | - | 4, 5 | 505 | |||
| AK150370.1 | - | 4, 5 | 504 | |||
| AK151324.1 | - | 4, 5 | 504 | |||
| AK151480.1 | - | 4, 5 | 504 | |||
| AK152477.1 | - | 4, 5 | 504 | |||
| AK152906.1 | - | 4, 5 | 504 | |||
| AK153068.1 | - | 4, 5 | 504 | |||
| AK153166.1 | - | 4, 5 | 504 | |||
| AK153395.1 | - | 4, 5 | 504 | |||
| AK154662.1 | - | 4, 5 | 504 | |||
| AK156481.1 | - | 4, 5 | 504 | |||
| AK157147.1 | - | 4, 5 | 504 | |||
| AK157170.1 | - | 4, 5 | 504 | |||
| AK157249.1 | - | 4, 5 | 504 | |||
| AK157700.1 | - | 4, 5 | 503 | |||
| AK159835.1 | - | 4, 5 | 504 | |||
| BC005450.1 | - | 4, 5 | 485 | 3487214 | ||
| CT010225.1 | - | 3, 4 | 450 | |||
| M32489.1 | - | 4, 5 | 470 | |||

Irf8
Genomic Map