siRNA Details
| Gene Symbol: Ier2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010499.4 | NP_034629.3 | 1 | 483 | ||
| GenBank mRNA | AK032692.1 | - | 481 | |||
| AK144601.1 | - | 1 | 480 | |||
| AK150860.1 | - | 1 | 478 | |||
| AK152747.1 | - | 1 | 478 | |||
| AK170433.1 | - | 480 | ||||
| BC002067.1 | - | 1 | 468 | 3590296 | ||
| BU604538.1 | - | 1 | 486 | |||

Ier2
Genomic Map