siRNA Details
| Gene Symbol: Ier2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010499.4 | NP_034629.3 | 1 | 389 | ||
| GenBank mRNA | AK032692.1 | - | 387 | |||
| AK144601.1 | - | 1 | 386 | |||
| AK150860.1 | - | 1 | 384 | |||
| AK152747.1 | - | 1 | 384 | |||
| AK170433.1 | - | 386 | ||||
| AK172332.1 | - | 1 | 332 | |||
| BC002067.1 | - | 1 | 374 | 3590296 | ||
| BU604538.1 | - | 1 | 392 | |||
| M31042.1 | - | 1 | 390 | |||
| M59821.1 | - | 1 | 391 | |||

Ier2
Genomic Map