siRNA Details
Gene Symbol: Jchain | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152839.3 | NP_690052.2 | 2 | 204 | ||
GenBank mRNA | AB644392.1 | - | 2 | 86 | ||
AK078865.1 | - | 2 | 90 | |||
AK160344.1 | - | 2 | 156 | |||
BC006026.1 | - | 2 | 108 | 3491646 | ||
J00544.1 | - | 2 | 91 | |||
M90766.1 | - | 2 | 107 |
