siRNA Details
| Gene Symbol: Il2ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008367.3 | NP_032393.3 | 3 | 474 | ||
| GenBank mRNA | AK037369.1 | - | 3 | 460 | ||
| AK088594.1 | - | 3 | 416 | |||
| AK171665.1 | - | 3 | 411 | |||
| BC114437.1 | - | 3 | 380 | 40054387 | ||
| K02891.1 | - | 3 | 357 | |||
| M30856.1 | - | 3 | 404 | |||
| M54934.1 | - | 3 | 277 | |||

Il2ra
Genomic Map