siRNA Details
| Gene Symbol: Il6st | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010560.3 | NP_034690.3 | 17 | 2850 | ||
| GenBank mRNA | AK142637.1 | - | 7 | 1311 | ||
| BC058679.1 | - | 17 | 2811 | 6834623 | ||
| M83336.1 | - | 16 | 2655 | |||
| X62646.1 | - | 16 | 2655 | |||

Il6st
Genomic Map