siRNA Details
| Gene Symbol: Gimap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008376.3 | NP_032402.2 | 3 | 962 | ||
| NM_175860.2 | NP_787056.1 | 3 | 956 | |||
| GenBank mRNA | AK156732.1 | - | 3 | 957 | ||
| AK169133.1 | - | 3 | 915 | |||
| BC132601.1 | - | 3 | 922 | 40130878 | ||
| BC132603.1 | - | 3 | 922 | 40130880 | ||
| Y08026.2 | - | 1 | 1071 | |||

Gimap1
Genomic Map