siRNA Details
| Gene Symbol: Inhba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008380.1 | NP_032406.1 | 3 | 784 | ||
| GenBank mRNA | AK134991.1 | - | 5 | 1235 | ||
| AK135474.1 | - | 2 | 616 | |||
| BC053527.1 | - | 3 | 777 | 6528822 | ||
| CT010380.1 | - | 2 | 612 | |||
| X69619.1 | - | 3 | 784 | |||

Inhba
Genomic Map