siRNA Details
| Gene Symbol: Inhbb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008381.3 | NP_032407.1 | 2 | 1655 | ||
| GenBank mRNA | AK132352.1 | - | 2 | 1657 | ||
| BC048845.1 | - | 2 | 361 | |||
| X83376.1 | - | 1 | 26 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Inhbb| Gene Symbol: Inhbb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008381.3 | NP_032407.1 | 2 | 1655 | ||
| GenBank mRNA | AK132352.1 | - | 2 | 1657 | ||
| BC048845.1 | - | 2 | 361 | |||
| X83376.1 | - | 1 | 26 | |||