siRNA Details
| Gene Symbol: Kcna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010595.3 | NP_034725.3 | 2 | 2859 | ||
| GenBank mRNA | AK039559.1 | - | 2 | 1854 | ||
| AK158065.1 | - | 2 | 1827 | |||
| BC112970.2 | - | 2 | 1464 | 40048563 | ||
| Y00305.1 | - | 1 | 2238 | |||

Kcna1
Genomic Map