siRNA Details
Gene Symbol: Kcnc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290682.1 | NP_001277611.1 | 2 | 1263 | ||
NM_008422.2 | NP_032448.2 | 2 | 1263 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kcnc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290682.1 | NP_001277611.1 | 2 | 1263 | ||
NM_008422.2 | NP_032448.2 | 2 | 1263 |