siRNA Details
| Gene Symbol: Kcnc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290682.1 | NP_001277611.1 | 2 | 1045 | ||
| NM_008422.2 | NP_032448.2 | 2 | 1045 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnc3| Gene Symbol: Kcnc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290682.1 | NP_001277611.1 | 2 | 1045 | ||
| NM_008422.2 | NP_032448.2 | 2 | 1045 | |||